Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylhydroquinone, 98%

Catalog No. p-7026680
Click to view available options
Quantity:
500 g

CAS: 700-13-0 | C9H12O2 | 152.193 g/mol

Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.

Solubility
Soluble in water (2 g/L @ 20°C.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-13-0
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002346
InChI Key AUFZRCJENRSRLY-UHFFFAOYSA-N
Synonym trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
PubChem CID 12785
IUPAC Name 2,3,5-trimethylbenzene-1,4-diol
SMILES CC1=CC(=C(C(=C1O)C)C)O

Specifications

Melting Point 169°C to 174°C
Flash Point 140°C (284°F)
Quantity 500 g
UN Number UN3077
Beilstein 1909183
Solubility Information Soluble in water (2g/L @ 20°C.
Formula Weight 152.2
Percent Purity 98%
Chemical Name or Material Trimethylhydroquinone
Hazard Category H310-H315-H317-H318-H332-H335
Hazard Statement GHS H Statement
H318-H332-H317-H335-H315
Causes serious eye damage.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P262-P264b-P270-P271-P272-P280-P302+P350-P304+P340-P305+P351+P338-P310-P312-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 211-838-3
RTECSNumber MX7820000
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.