Alfa Aesar™ Trimethylphosphine, 98%

Manufacturer: Alfa Aesar™ 3014314

Quantity 25g

 View more versions of this product

Catalog No. AA3014314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 594-09-2
    Formula Weight 76.08
    MDL Number MFCD00008510
    UN Number UN2845
    Synonym Methyl phosphine
    Chemical Name or Material Trimethylphosphine

    Specifications

    Physical Form Liquid, Ampouled Under Argon
    Formula Weight 76.08
    Health Hazard 2 GHS H Statement
    H225-H250-H252-H314-H318
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2845
    Quantity 25g
    Odor Pungent
    MDL Number MFCD00008510
    CAS 594-09-2
    Synonym Methyl phosphine
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Density 0.735g/mL
    Flash Point −30° (−22°F)
    Sensitivity Air & Moisture Sensitive
    Boiling Point 38° to 40°C
    Chemical Name or Material Trimethylphosphine
    Assay 99%

    GHS H Statement
    H225-H250-H252-H314-H318
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit