Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylphosphine oxide, Thermo Scientific Chemicals

Catalog Number p-7049861
Click to view available options
Quantity:
1 g
5 g
676-96-0
C3H9OP
92.078
MFCD00013910
LRMLWYXJORUTBG-UHFFFAOYSA-N
trimethylphosphine oxide, phosphine oxide, trimethyl, ch3 3po, phosphine oxide,trimethyl-6ci,8ci,9ci, dimethylphosphoryl methane, trimethylphosphane oxide, trimethylphosphineoxide, me3po, acmc-1bhzf, trimethyloxylatophosphonium
69609
dimethylphosphorylmethane
CP(=O)(C)C
676-96-0
C3H9OP
92.078
MFCD00013910
LRMLWYXJORUTBG-UHFFFAOYSA-N
trimethylphosphine oxide, phosphine oxide, trimethyl, ch3 3po, phosphine oxide,trimethyl-6ci,8ci,9ci, dimethylphosphoryl methane, trimethylphosphane oxide, trimethylphosphineoxide, me3po, acmc-1bhzf, trimethyloxylatophosphonium
69609
dimethylphosphorylmethane
CP(=O)(C)C

Trimethylphosphine oxide is a common side product in organic synthesis, including the Wittig, Staudinger, Mitsunobu reactions and also in the manufacture of triphenylphosphine. It is a popular reagent to induce the crystallization of various chemical compounds. It acts as an excellent ligand for hard metal centers. It can react with 9-methyl-9-azonianthracene methyl sulfate to get (10-methyl-9,10-dihydro-acridin-9-yl)-phosphonic acid dimethyl ester using sodium iodide and acetonitrile as solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 140°C to 141°C
Odor Odorless
Quantity 1 g
Solubility Information Soluble in polar organic solvents.
Formula Weight 92.08
Physical Form Crystalline
Chemical Name or Material Trimethylphosphine oxide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-633-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit