Alfa Aesar™ Trimethylpyruvic acid, ca. 60% aq. soln.

Manufacturer: Alfa Aesar™ B2558218

Quantity 50g

 View more versions of this product

Catalog No. AAB2558218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 815-17-8
    Molecular Formula C6H10O3
    Linear Formula (CH3)3CCOCO2H
    Formula Weight 130.14
    MDL Number MFCD00154352
    UN Number UN3265
    Synonym 3,3-Dimethyl-2-oxobutyric acid
    Chemical Name or Material Trimethylpyruvic acid

    Specifications

    Formula Weight 130.14
    Linear Formula (CH3)3CCOCO2H
    Synonym 3,3-Dimethyl-2-oxobutyric acid
    Concentration ≈60% aq. Soln.
    Quantity 50g
    TSCA TSCA
    Molecular Formula C6H10O3
    MDL Number MFCD00154352
    CAS 815-17-8
    UN Number UN3265
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Trimethylpyruvic acid

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit