Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trimethylpyruvic acid, ca. 60% aq. soln.

Click to view available options
Quantity:
50g
250g
815-17-8
C6H10O3
130.143
MFCD00154352
IAWVHZJZHDSEOC-UHFFFAOYSA-N
3,3-dimethyl-2-oxobutyric acid,trimethylpyruvic acid,butanoic acid, 3,3-dimethyl-2-oxo,pyruvic acid, trimethyl,glyoxylic acid, tert-butyl,unii-29cl615clh,3,3-dimethyl-2-oxo-butanoic acid,butyric acid, 3,3-dimethyl-2-oxo,tert-butylglyoxylic acid,trimethlpyruvic acid
13150
3,3-dimethyl-2-oxobutanoic acid
CC(C)(C)C(=O)C(=O)O
815-17-8
C6H10O3
130.143
MFCD00154352
IAWVHZJZHDSEOC-UHFFFAOYSA-N
3,3-dimethyl-2-oxobutyric acid,trimethylpyruvic acid,butanoic acid, 3,3-dimethyl-2-oxo,pyruvic acid, trimethyl,glyoxylic acid, tert-butyl,unii-29cl615clh,3,3-dimethyl-2-oxo-butanoic acid,butyric acid, 3,3-dimethyl-2-oxo,tert-butylglyoxylic acid,trimethlpyruvic acid
13150
3,3-dimethyl-2-oxobutanoic acid
CC(C)(C)C(=O)C(=O)O
Quantity 250g
Linear Formula (CH3)3CCOCO2H
UN Number UN3265
Concentration or Composition (by Analyte or Components) ≈60% aq. Soln.
Formula Weight 130.14
Chemical Name or Material Trimethylpyruvic acid

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA