Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 94%

Manufacturer:  Alfa Aesar™ H2685703

Quantity 1g

 View more versions of this product

Catalog No. AAH2685703


  Check Availability
Add to cart

Specifications

Description

Applications
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

Solubility
Reacts with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

120801-75-4
250.27
UN2920
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
FSO2CF2CO2Si(CH3)3
MFCD02093343
2,2-Difluoro-2-(fluorosulfonyl)acetic acid trimethylsilyl ester

Specifications

26°C (79°F)
1g
FSO2CF2CO2Si(CH3)3
MFCD02093343
2,2-Difluoro-2-(fluorosulfonyl)acetic acid trimethylsilyl ester
94%
156°C
120801-75-4
250.27
UN2920
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit