Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trimethylsilyl acetate, 97%

Click to view available options
Quantity:
25g
100g
2754-27-0
C5H12O2Si
132.234
MFCD00008695
QHUNJMXHQHHWQP-UHFFFAOYSA-N
acetoxytrimethylsilane,trimethylacetoxysilane,acetato trimethylsilane,acetic acid trimethylsilyl ester,silanol, trimethyl-, acetate,silanol, 1,1,1-trimethyl-, 1-acetate,tmsoac,o-trimethylsilylacetate,trimethylsilyl acetate
75988
trimethylsilyl acetate
CC(=O)O[Si](C)(C)C
2754-27-0
C5H12O2Si
132.234
MFCD00008695
QHUNJMXHQHHWQP-UHFFFAOYSA-N
acetoxytrimethylsilane,trimethylacetoxysilane,acetato trimethylsilane,acetic acid trimethylsilyl ester,silanol, trimethyl-, acetate,silanol, 1,1,1-trimethyl-, 1-acetate,tmsoac,o-trimethylsilylacetate,trimethylsilyl acetate
75988
trimethylsilyl acetate
CC(=O)O[Si](C)(C)C
Boiling Point 107°C
Flash Point 4°C (39°F)
Quantity 100g
Melting Point -32°C
Linear Formula CH3CO2Si(CH3)3
UN Number UN3272
Beilstein 1744463
Formula Weight 132.24
Percent Purity 97%
Chemical Name or Material Trimethylsilyl acetate

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA