Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trimethylsilyl acetate, 97%

Manufacturer:  Alfa Aesar™ B2203714

Quantity 25g

 View more versions of this product

Catalog No. AAB2203714


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2754-27-0
CH3CO2Si(CH3)3
MFCD00008695
1744463
Trimethylsilyl acetate
C5H12O2Si
132.24
UN3272
Acetic acid trimethylsilyl ester; Acetoxytrimethylsilane

Specifications

4°C (39°F)
−32°C
2754-27-0
CH3CO2Si(CH3)3
MFCD00008695
1744463
Trimethylsilyl acetate
107°C
25g
C5H12O2Si
132.24
UN3272
Acetic acid trimethylsilyl ester; Acetoxytrimethylsilane
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit