Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl cyanide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
500g
Packaging:
Glass bottle
7677-24-9
C4H9NSi
99.208
MFCD00001765
LEIMLDGFXIOXMT-UHFFFAOYSA-N
trimethylsilyl cyanide, cyanotrimethylsilane, trimethylsilanecarbonitrile, trimethylsilylcyanide, silanecarbonitrile, trimethyl, trimethylsilylnitrile, tmscn, trimethyl silane cyanide, trimethylsilylcarbonitrile, tms cyanide
82115
trimethylsilylformonitrile
C[Si](C)(C)C#N
7677-24-9
C4H9NSi
99.208
MFCD00001765
LEIMLDGFXIOXMT-UHFFFAOYSA-N
trimethylsilyl cyanide, cyanotrimethylsilane, trimethylsilanecarbonitrile, trimethylsilylcyanide, silanecarbonitrile, trimethyl, trimethylsilylnitrile, tmscn, trimethyl silane cyanide, trimethylsilylcarbonitrile, tms cyanide
82115
trimethylsilylformonitrile
C[Si](C)(C)C#N
C-X bond formation reagent
Assay Percent Range 98%
Linear Formula (CH3)3SiCN
Fieser 04,542; 05,720; 06,632; 07,397; 08,133; 09,127; 10,112; 11,147; 12,148; 13,87; 14,107; 15,102; 17,89; 01,979; 04,418; 06,503; 08,433
Formula Weight 99.21
Packaging Glass bottle
Quantity 25g
Chemical Name or Material Trimethylsilyl cyanide

GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.

GHS P Statement:
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger