Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (Trimethylsilyl)diazomethane, 2M in hexanes

Manufacturer:  Alfa Aesar™ H26744AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH26744AA


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

Applications
(Trimethylsilyl)diazomethane is used as a methylating agent in organic chemistry. It is utilized in Gas Chromatography- Mass-Spectrometry (GC-MS) for the analysis of various carboxylic compounds. It is involved as a reagent in the Doyle-Kirmse reaction with allyl sulfides and allyl amines. It is considered and used as a less-explosive replacement fordiazomethane. It is also used as a reactant for Unnatural alfa-amino acid derivatives containing gem-bisphosphonates through Michael addition reaction, imidazolo[2,1-b]benzothiazole derivatives and aigialomycin D analogues.

Solubility
Immiscible with water. Miscible with organic solvents.

Notes
(Trimethylsilyl)diazomethane (TMSD) in alcoholic solvents under acidic or basic conditions can lead to the formation of diazomethane. TMSD is under review by the National Toxicological Program for potential inhalation hazards.Incompatible with oxidizing agents, chlorine, fluorine and perchlorates.
Description & Specifications

Chemical Identifiers

18107-18-1
114.22
UN3383
(Diazomethyl)trimethylsilane
C4H10N2Si
MFCD00053946
1902903
(Trimethylsilyl)diazomethane

Specifications

18107-18-1
114.22
UN3383
(Diazomethyl)trimethylsilane
5mL
(Trimethylsilyl)diazomethane
C4H10N2Si
MFCD00053946
1902903
2M in hexanes
−23°C (−10°F)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H331-H350-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
Toxic if inhaled.
May cause cancer.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit