Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trimethylsilyl)diazomethane, approx. 2M solution in diethyleether, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
18107-18-1,60-29-7
C4H10N2Si
114.223
MFCD00053946
ONDSBJMLAHVLMI-UHFFFAOYSA-N
trimethylsilyl diazomethane,trimethylsilyldiazomethane,diazomethyl trimethylsilane,tms-diazomethane,diazomethyl trimethyl silane,tmschn2,silane, diazomethyl trimethyl,unii-qi98hqo8c4,diazomethyl-trimethyl-silane,qi98hqo8c4
167693
diazomethyl(trimethyl)silane
C[Si](C)(C)C=[N+]=[N-]
18107-18-1,60-29-7
C4H10N2Si
114.223
MFCD00053946
ONDSBJMLAHVLMI-UHFFFAOYSA-N
trimethylsilyl diazomethane,trimethylsilyldiazomethane,diazomethyl trimethylsilane,tms-diazomethane,diazomethyl trimethyl silane,tmschn2,silane, diazomethyl trimethyl,unii-qi98hqo8c4,diazomethyl-trimethyl-silane,qi98hqo8c4
167693
diazomethyl(trimethyl)silane
C[Si](C)(C)C=[N+]=[N-]
Alkylation reagent
Quantity 5mL
Packaging Glass bottle
Flash Point −35°C
Color White
Specific Gravity 0.77
Density 0.7700g/mL
CAS Min % 28.19,67.36
CAS Max % 32.64,71.81
Formula Weight 114.23
Physical Form Crystals or Crystalline Powder
Chemical Name or Material (Trimethylsilyl)diazomethane, Approx. 2M solution in diethyl ether
Show More Show Less

GHS H Statement
May cause drowsiness or dizziness. May cause cancer. Harmful if swallowed. Fatal if inhaled. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Obtain special instructions before use. Do not breathe dust/fume/gas/mist/vapors/spray. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Wash face, hands and any exposed skin thoroughly after handling.

Danger