Learn More
Trimethylsilyl isocyanate, 94%, Thermo Scientific Chemicals
Description
Trimethylsilyl isocyanate is used in the preparation of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones. It finds application in the carbamoylation of aromatic hydrocarbons and alcohols. It is also used in the preparation of primary amides from Grignard reagents. Further, it is used in the synthesis of cyclic imides. It is also utilized for the synthesis of trifluoroacetyl isocyanate and cyclic imides. In addition to this, it is employed in the conversion of isocyanates to carbodiimides using cyclopentadienyl Mn(CO)3 catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trimethylsilyl isocyanate is used in the preparation of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones. It finds application in the carbamoylation of aromatic hydrocarbons and alcohols. It is also used in the preparation of primary amides from Grignard reagents. Further, it is used in the synthesis of cyclic imides. It is also utilized for the synthesis of trifluoroacetyl isocyanate and cyclic imides. In addition to this, it is employed in the conversion of isocyanates to carbodiimides using cyclopentadienyl Mn(CO)3 catalysis.
Solubility
Miscible with ethers and o-dichlorobenzene.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, strong bases, alcohols and amines.
Specifications
Melting Point | -49°C |
Density | 0.859 |
Boiling Point | 90°C to 92°C |
Flash Point | −2°C (28°F) |
Odor | Acrid |
Refractive Index | 1.396 |
Quantity | 10 g |
UN Number | UN2478 |
Beilstein | 1744962 |
Sensitivity | Moisture sensitive; May form a precipitate |
Show More |
Safety and Handling
Hazard Category | H225-H315-H319-H331-H334-H335 |
Hazard Statement | GHS H Statement H225-H330-H334-H315-H319-H335 Highly flammable liquid and vapour. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P311-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOCYANATES, FLAMMABLE, TOXIC, N.O.S. |
EINECSNumber | 214-256-8 |
TSCA | Yes |
Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.