Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
1822-00-0,92112-69-1,92112-69-1
C4H11LiSi
94.16
MFCD00010747
KVWLUDFGXDFFON-UHFFFAOYSA-N
trimethylsilyl methyllithium,trimethylsilyl methyl lithium,lich2sime3,pubchem18144,tms-ch2li,lithiomethyltrimethylsilane,acmc-1bvf0,trimethylsilylmethyl lithium
3482579
lithium;methanidyl(trimethyl)silane
[Li+].C[Si](C)(C)[CH2-]
1822-00-0,92112-69-1,92112-69-1
C4H11LiSi
94.16
MFCD00010747
KVWLUDFGXDFFON-UHFFFAOYSA-N
trimethylsilyl methyllithium,trimethylsilyl methyl lithium,lich2sime3,pubchem18144,tms-ch2li,lithiomethyltrimethylsilane,acmc-1bvf0,trimethylsilylmethyl lithium
3482579
lithium;methanidyl(trimethyl)silane
[Li+].C[Si](C)(C)[CH2-]
Alkylation reagent
Packaging AcroSeal™ Glass bottle
Color Brown-Yellow to Colorless
Quantity 100mL
Specific Gravity 0.65
Density 0.6500g/mL
Flash Point −40°C
Boiling Point 65.0°C to 70.0°C
CAS Min % 7.0,86.0
CAS Max % 14.0,93.0
Linear Formula LiCH2Si(CH3)3
Formula Weight 94.16
Physical Form Solution
Percent Purity 7 to 14 wt%
Chemical Name or Material (Trimethylsilyl)methyllithium, 0.8M (12 wt%) solution in hexanes
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
May cause d

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger