Learn More
(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 377458000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
(Trimethylsilyl)methyllithium | |
1822-00-0, 92112-69-1, 92112-69-1 | |
14.0, 93.0 | |
0.6500g/mL | |
−40°C | |
C4H11LiSi | |
800 mL | |
0.65 | |
KVWLUDFGXDFFON-UHFFFAOYSA-N | |
lithium;methanidyl(trimethyl)silane | |
3482579 | |
7 to 14 wt% |
0.8M (12 wt%) solution in hexanes | |
7.0, 86.0 | |
Brown-Yellow to Colorless | |
65.0°C to 70.0°C | |
AcroSeal glass Bottle packed in tin can | |
LiCH2Si(CH3)3 | |
MFCD00010747 | |
trimethylsilyl methyllithium, trimethylsilyl methyl lithium, lich2sime3, pubchem18144, tms-ch2li, lithiomethyltrimethylsilane, acmc-1bvf0, trimethylsilylmethyl lithium | |
[Li+].C[Si](C)(C)[CH2-] | |
94.16 | |
94.16 | |
Solution |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
May cause d
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only