Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl trifluoroacetate, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
400-53-3
C5H9F3O2Si
186.21
MFCD00000413
VIYXXANHGYSBLY-UHFFFAOYSA-N
trimethylsilyl trifluoroacetate,trimethylsilyltrifluoroacetate,acetic acid, trifluoro-, trimethylsilyl ester,acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester,trifluoroacetic acid trimethylsilyl ester,tms trifluoroacetate,acmc-1ago5,cf3c o osi ch3 3,trimethylsilyl-trifluoroacetate,trifluoroacetic acid, tms derivative
67863
trimethylsilyl 2,2,2-trifluoroacetate
C[Si](C)(C)OC(=O)C(F)(F)F
400-53-3
C5H9F3O2Si
186.21
MFCD00000413
VIYXXANHGYSBLY-UHFFFAOYSA-N
trimethylsilyl trifluoroacetate,trimethylsilyltrifluoroacetate,acetic acid, trifluoro-, trimethylsilyl ester,acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester,trifluoroacetic acid trimethylsilyl ester,tms trifluoroacetate,acmc-1ago5,cf3c o osi ch3 3,trimethylsilyl-trifluoroacetate,trifluoroacetic acid, tms derivative
67863
trimethylsilyl 2,2,2-trifluoroacetate
C[Si](C)(C)OC(=O)C(F)(F)F
Boiling Point 88.0°C to 90.0°C
Flash Point 0°C
Color White
Infrared Spectrum Authentic
Specific Gravity 1.08
Quantity 25g
Refractive Index 1.3350 to 1.3380
Packaging Glass bottle
Density 1.0800g/mL
Assay Percent Range 95% min. (GC)
Linear Formula CF3CO2Si(CH3)3
Beilstein 04,IV,4004
Solubility Information Solubility in water: reacts
Formula Weight 186.21
Physical Form Powder
Percent Purity 96%
Chemical Name or Material Trimethylsilyl trifluoroacetate, 96%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr

GHS Signal Word: Danger

EINECSNumber : 206-923-7

TSCA : TSCA