Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethylsilyl trifluoroacetate, 96%, ACROS Organics™
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
Specifications
Specific Gravity | 1.08 |
Refractive Index | 1.3350 to 1.3380 |
Packaging | Glass bottle |
Quantity | 100g |
Flash Point | 0°C |
Infrared Spectrum | Authentic |
Density | 1.0800g/mL |
Boiling Point | 88.0°C to 90.0°C |
Color | White |
Assay Percent Range | 95% min. (GC) |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr
GHS Signal Word: Danger
EINECSNumber : 206-923-7
TSCA : TSCA