Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsulfonium Iodide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
2181-42-2
C3H9IS
204.07
MFCD00011632
VFJYIHQDILEQNR-UHFFFAOYSA-M
trimethylsulfonium iodide,trimethylsulphonium iodide,sulfonium, trimethyl-, iodide,trimethylsulfoniumiodide,trimethylsulfanium iodide,trimethyl-sulfonium iodide,unii-4z1liz2lun,4z1liz2lun,sulfonium, trimethyl-, iodide 1:1,chembl25340
75127
trimethylsulfanium;iodide
C[S+](C)C.[I-]
2181-42-2
C3H9IS
204.07
MFCD00011632
VFJYIHQDILEQNR-UHFFFAOYSA-M
trimethylsulfonium iodide,trimethylsulphonium iodide,sulfonium, trimethyl-, iodide,trimethylsulfoniumiodide,trimethylsulfanium iodide,trimethyl-sulfonium iodide,unii-4z1liz2lun,4z1liz2lun,sulfonium, trimethyl-, iodide 1:1,chembl25340
75127
trimethylsulfanium;iodide
C[S+](C)C.[I-]
Packaging Plastic bottle
Infrared Spectrum Authentic
Melting Point 215.0°C to 220.0°C
Quantity 500g
Color White to Yellow
Assay Percent Range 98%
Linear Formula (CH3)3SI
Fieser 01,314; 02,169; 01,1236; 02,171; 13,15
Solubility Information Solubility in water: soluble
Formula Weight 204.07
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material Trimethylsulfonium iodide
Show More Show Less