Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsulfonium methylsulfate, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2181-44-4
C4H12O4S2
188.27
MFCD00012205
ANXKZXRDXAZQJT-UHFFFAOYSA-M
trimethylsulfonium methyl sulfate,trimethylsulfonium methylsulfate,methyl sulfate; trimethylsulfanium,trimethylsulfonium methyl sulfate 1-,acmc-20ajcy,trimethylsulphonium methylsulphate,trimethylsulfonium methyl sulphate,methyl sulfate; trimethylsulfonium,trimethyl sulfonium methyl sulfate,trimethylsulphonium methyl-sulphate
2760871
methyl sulfate;trimethylsulfanium
COS(=O)(=O)[O-].C[S+](C)C
2181-44-4
C4H12O4S2
188.27
MFCD00012205
ANXKZXRDXAZQJT-UHFFFAOYSA-M
trimethylsulfonium methyl sulfate,trimethylsulfonium methylsulfate,methyl sulfate; trimethylsulfanium,trimethylsulfonium methyl sulfate 1-,acmc-20ajcy,trimethylsulphonium methylsulphate,trimethylsulfonium methyl sulphate,methyl sulfate; trimethylsulfonium,trimethyl sulfonium methyl sulfate,trimethylsulphonium methyl-sulphate
2760871
methyl sulfate;trimethylsulfanium
COS(=O)(=O)[O-].C[S+](C)C
Alkylation reagent
Packaging Glass bottle
Color White
Melting Point 92.0°C to 94.0°C
NMR 97.5% min.
Quantity 25g
Assay Percent Range 98%
Formula Weight 188.27
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Trimethylsulfonium methylsulfate

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning