Alfa Aesar™ Trimethyltin chloride

Manufacturer: Alfa Aesar™ 7116618

Quantity 50g

 View more versions of this product

Catalog No. AA7116618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1066-45-1
    Molecular Formula (CH3)3SnCl
    Formula Weight 199.25
    MDL Number MFCD00000520
    UN Number UN3146
    Synonym Chlorotrimethyltin
    Chemical Name or Material Trimethyltin chloride

    Specifications

    Formula Weight 199.25
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 50g
    Boiling Point 154°C
    Physical Form Crystalline
    CAS 1066-45-1
    Health Hazard 2 GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    RTECS Number WH6850000
    TSCA TSCA
    UN Number UN3146
    Color White
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    Sensitivity Moisture Sensitive
    Melting Point 37° to 39°C
    Molecular Formula (CH3)3SnCl
    MDL Number MFCD00000520
    Synonym Chlorotrimethyltin
    Flash Point 97°C (207°F)
    Health Hazard 1 Danger
    Odor Unpleasant
    Chemical Name or Material Trimethyltin chloride

    GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit