Alfa Aesar™ Trimethylurea

Manufacturer: Alfa Aesar™ 1647503

Quantity 1g

 View more versions of this product

Catalog No. AA1647503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 632-14-4
    Formula Weight 102.14
    MDL Number MFCD00014868
    Chemical Name or Material Trimethylurea

    Specifications

    Physical Form Solid/Semi-Solid Mass
    Formula Weight 102.14
    Odor Characteristic
    MDL Number MFCD00014868
    Melting Point 74° to 75°C
    Quantity 1g
    CAS 632-14-4
    Sensitivity Hygroscopic
    Chemical Name or Material Trimethylurea

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit