Learn More
Aviva Systems Biology Corp TRIML2 Antibody - middle region (ARP55636_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55636_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIML2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIML2. Alternate names: FLJ25801, MGC138164, MGC138166, spryd6, SPRYD6ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61562