Learn More
Mopec Large Sink Countertop Trimming Station
Designed to maximize floor space while allowing for convenient handling of large specimens. Mopec™ Large Sink Countertop trimming Station offers limitless utility for the smallest to even the largest labs.
Manufacturer: Mopec OB200
Description
- All stainless steel construction
- Includes Hot/cold water fixture
Wrist blade handles with swing spout faucet and built-in vacuum breaker - Recessed stainless steel work surface with removable polyethylene cutting board, 58.4 x 40.6 x 2cm
- One large and one small stainless steel organizer bin
- Washable rule with inch and centimeter markings
- Ruler is located on the inside lip of the workstation
- Magnetic tool bar holds instruments when not in use
- Stainless steel C-Fold paper towel dispenser
- Two energy saving, long life LED lights
- Back draft ventilation
- Designed to safely and efficiently remove formaldehyde fumes and hazardous chemical vapors
- Equipped with a factory installed exhaust duct which can be connected to any in-house ventilation system
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
1 Year Warranty
Specifications
48 in. | |
121.9cm | |
GFCI electrical receptacle | |
115.6cm | |
116.9 x 7.6cm duct stub connection | |
48cm | |
115V | |
14 in. | |
36cm |
10 in. | |
25cm | |
45.5cm | |
60HZ | |
19 in. | |
Trimming Station | |
29 in. | |
73.7cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.