Nova Biostorage Trinean DropPlate 96-S
Microfluidic plates that house samples for analysis within the Trinean DropSense 96 system
Manufacturer: Nova Biostorage MT20012BX
 View all options for this product
Catalog No. 50-104-7462
  Check Availability
Add to Cart
    Includes Box of 10
    Item Description DropPlate 96-S
    • 2µL sample required: minimal sample consumption
    • 16-well or 96-well DropPlate formats: fit for low to high throughput
    • Microplate dimensions: automation friendly
    • Barcoded DropPlate 96: ideal for sample tracking
    • Capillary storage channel: easy loading, protection from evaporation or cross-contamination
    • Compatible with viscous or concentrated samples: fit for all kinds of biomolecules
    Two types of DropPlates available:
    • DropPlate S: Microfluidic plate with a single measurement chamber, ideal for routine genomic applications (DNA and RNA extractions or PCR amplicon quantification).
    • DropPlate D+: Microfluidic plate with a dual measurement chamber for an extended OD range for broad range applications (from sensitive NGS sample quantification to highly concentrated proteins or oligonucleotides QC).
  • Description & Specifications
Related Products
Product Content Feedback