Learn More
Trinity Biotech A1c Analyzer
Interference-free A1c results in two minutes
Manufacturer:  Trinity Biotech, Inc 03010050
 View more versions of this product
Catalog No. 22-140-142
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
ASTM 1384
Liquid chromatograph (high pressure pump, column oven and fixed wavelength detector), 10μL injection valve, printer, and pre-programmed GHb/A1c PDQ system software
2.33 min./Injection
A1c analyzer standalone