Trinity Biotech™ HbA1c Reagents
Specially formulated for PDQ/PDQ Plus Systems
Manufacturer: Trinity Biotech, Inc 01030065
 View more versions of this product
Catalog No. 22-140-145
  Check Availability
Add to Cart
Disclaimers:
For in vitro diagnostic use only.
    Specifications
    Product Type PDQ2 Buffer reagent
    For Use With (Application) Quantitative measurement of the percentage of total glycated hemoglobins in whole Blood, in-vitro diagnostic use only
    For Use With (Equipment) PDQ, PDQ Plus system
    Quantity 940mL
    Certifications/Compliance CE Marked, FDA
  • Description & Specifications
Related Products
Product Content Feedback