Trinity Biotech™ Moblie Phase Reagent

Designed for use with the Trinity Biotech ultra2 systems

Manufacturer: Trinity Biotech, Inc 01030041

 View more versions of this product

Catalog No. 22-140-162

  Check Availability
Add to Cart

    Specifications

    Product Type Mobile phase 2 reagent
    Format Liquid
    For Use With (Application) In-vitro diagnostic use only
    For Use With (Equipment) Ultra2 system
    Certifications/Compliance CE, FDA
    Storage Requirements 15° to 28°C
    Quantity 3800mL,3800mL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit