Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trinity Biotech Premier Hb9210™ - HbA1c Analyzer

With proprietary boronate affinity technology, this analyzer offers accurate and precise results, free from interference, in a rapid and highly scalable package. Trinity Biotech Premier Hb9210 uses the glycation specific binding of boronate affinity to detect all of the glycated Hb species present.

Manufacturer:  Trinity Biotech 09000001

Catalog No. 22-140-171


Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

  • Consists of an integrated HPLC system, a compact sample handler and the workstation with Affinity™ control software
  • Touch-screen user-interface
  • Sample analysis time of just 66 seconds, delivers a throughput of 54 tests per hour
  • Results need minimal interpretation; CV below 2%
  • Capacity: 210 patient samples
  • Racked tubes allows operator to quickly add samples, prior to the instrument aspirating them directly from closed whole blood sample tubes using a self-cleaning syringe mechanism
  • Color- and bar-coded reagents with excellent onboard stability, electronic level sensing and a continuous display of reagent status ensures sufficient reagents are onboard at all times
  • Certified to report in the following units: IFCC (mMol HbA1c/mol Hb) and NGSP (% HbA1c)
  • Affinity software combines operator ease-of-use with the advanced data management facilities and LIS integration
  • Precision: Intra-run CVs below 2% guarantee a reliable, repeatable result
  • Linearity: From 3.8%HbA1c to 18.5%HbA1c ensures accuracy for the whole diabetic population
  • Specificity: minimal interference from variants, interferents, matrix effects and sample freezing
Specifications
  • Instrumentation Control: Windows Operating System with Proprietary Assay Software
  • Results Output: Display and Print
  • Printout: Automatic, User Select
  • Sample Types: Whole blood, hemolysates made from whole blood or packed red blood cells
  • Minimum sample requirement: 10μL (whole blood to make 1:150 hemolysate dilution); 0.5mL (whole blood to make direct injection)
  • Sample ID: Operator Input or Bar Code Reader
  • Calibration: Previous IFCC/NGSP calibration used for each sample
  • Dimensions (D x W x H): 66 x 54 x 74cm
  • Weight: 62kg

Certifications

FDA, CE

Specifications

Specifications

54cm
66cm
Detect all of the glycated Hb species present in blood sample
CE, FDA
10μL, 0.5mL
62kg
Integrated HPLC system, a compact sample handler and the workstation with Affinity™ control software
74cm
Premier Hb9210 - HbA1c Analyzer
Whole blood, hemolysates made from whole blood or packed red blood cells
66 seconds
HbA1c
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit