Trinity Biotech Uni-Gold™ Recombignen™ HIV-1/2 Available on GSA/VA Contract for Federal Government customers only.

Suitable for use in appropriate multi-test algorithms designed for the statistical validation of rapid HIV test results

Manufacturer: Trinity Biotech, Inc 1206530

 View more versions of this product

Catalog No. 22-140-211

  Check Availability
Add to Cart

    Specifications

    Product Type Uni-Gold™ Recombigen™ HIV-1/2 test kit control
    Detectable Analytes HIV-1 and HIV-2 Antibodies in Serum, Plasma and whole Blood
    For Use With (Equipment) Uni-Gold™ Recombigen™ HIV-1/2
    Includes Positive HIV-1 of 500μL, positive HIV-2 of 500μL, negative 500μL
    Quantity Each
    Sample Size 500μL
    Sensitivity 100%
    Specificity 99.7%
    Storage Requirements 2° to 8°C
    Clia Complexity Waived, moderate

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit