Learn More
Sklar InstrumentsSupplier Diversity Partner Trinket Box, Sklar, 4 in. x 2-3/4 in. x 1 in. Trinket Box Peforated Lid, 105mm x 70mm x 25mm. Sklar's Trinket Box is designed for use in washing and sterilizing small, loose parts of instruments. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106752
Trinket Box, Sklar, 4 in. x 2-3/4 in. x 1 in. Trinket Box Peforated Lid, 105mm x 70mm x 25mm. Sklar's Trinket Box is designed for use in washing and sterilizing small, loose parts of instruments. It is available in multiple sizes.
Catalog No. 50-143-7297