Learn More
Sklar InstrumentsSupplier Diversity Partner Trinket Box, Sklar, 5 in. x 3 in. x 1 in. Trinket Box Wire Mesh Lid, 120mm x 80mm x 25mm. Sklar's Trinket Box is designed for use in washing and sterilizing small, loose parts of instruments. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106764
Trinket Box, Sklar, 5 in. x 3 in. x 1 in. Trinket Box Wire Mesh Lid, 120mm x 80mm x 25mm. Sklar's Trinket Box is designed for use in washing and sterilizing small, loose parts of instruments. It is available in multiple sizes.
Catalog No. 50-143-7301