Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trioctylsilane, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
18765-09-8
C24H52Si
368.76
QTKHQYWRGFZFHG-UHFFFAOYSA-N
trioctylsilane,silane, trioctyl,tri-n-octylsilane,trioctylsilyl
6327367
trioctylsilicon
CCCCCCCC[Si](CCCCCCCC)CCCCCCCC
18765-09-8
C24H52Si
368.76
QTKHQYWRGFZFHG-UHFFFAOYSA-N
trioctylsilane,silane, trioctyl,tri-n-octylsilane,trioctylsilyl
6327367
trioctylsilicon
CCCCCCCC[Si](CCCCCCCC)CCCCCCCC
Reduction reagent
Specific Gravity 0.821
Color Colorless
Density 0.8210g/mL
Flash Point 113°C
Boiling Point 163.0°C to 165.0°C (0.2mmHg)
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Formula Weight 368.76
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Trioctylsilane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 242-559-5