Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triosephosphate isomerase Mouse anti-Human, Mouse, Rat, Biotin, Clone: 3711, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243825B

Catalog No. NBP243825B


Add to cart

Description

Description

Triosephosphate isomerase Monoclonal antibody specifically detects Triosephosphate isomerase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Triosephosphate isomerase
3711
Western Blot
Purified
TPI1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Triosephosphate isomerase. The exact sequence is proprietary.
Protein G purified
Stem Cell Markers
Primary
7167
Western Blot
Biotin
PBS with 0.05% Sodium Azide
EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit