Learn More
Triosephosphate isomerase Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: 1111, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243824UV
Catalog No. NBP243824UV
Add to cart
Description
Description
Triosephosphate isomerase Monoclonal antibody specifically detects Triosephosphate isomerase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
Triosephosphate isomerase
1111
Western Blot
Purified
TPI1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Triosephosphate isomerase. The exact sequence is proprietary.
Protein G purified
Stem Cell Markers
Primary
7167
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat