Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Triosephosphate isomerase Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: 3711, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243825FR
Catalog No. NBP243825FR
Description
Description
Triosephosphate isomerase Monoclonal antibody specifically detects Triosephosphate isomerase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
Triosephosphate isomerase
3711
Western Blot
Purified
TPI1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Triosephosphate isomerase. The exact sequence is proprietary.
Protein G purified
Stem Cell Markers
Primary
7167
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat