Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIP6 Mouse anti-Human, PE, Clone: 6H4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237440PE

Catalog No. NBP237440PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TRIP6
6H4
Flow Cytometry
Purified
TRIP6
Purified recombinant fragment of human TRIP6 (AA: 107-291) expressed in E. Coli.
Protein A or G purified
Core ESC Like Genes, Stem Cell Markers
Primary
7205
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
MGC10556, MGC10558, MGC29959, MGC3837, MGC4423, OIP1OIP-1, Opa-interacting protein 1, thyroid hormone receptor interactor 6, thyroid receptor-interacting protein 6, TR-interacting protein 6, ZRP-1TRIP-6, zyxin related protein 1, Zyxin-related protein 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit