Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripelennamine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
154-69-8
C16H22ClN3
291.82
FSSICIQKZGUEAE-UHFFFAOYSA-N
N-benzyl-N-[2-(dimethylamino)ethyl]pyridin-2-amine hydrochloride
Cl.CN(C)CCN(CC1=CC=CC=C1)C1=CC=CC=N1
154-69-8
C16H22ClN3
291.82
FSSICIQKZGUEAE-UHFFFAOYSA-N
N-benzyl-N-[2-(dimethylamino)ethyl]pyridin-2-amine hydrochloride
Cl.CN(C)CCN(CC1=CC=CC=C1)C1=CC=CC=N1

T1120, 154-69-8, C16H21N3•HCl

Spectrum™ Tripelennamine Hydrochloride, USP is an antihistamine used to treat sneezing, runny nose, itching, watery eyes, and other symptoms of allergies and the common cold. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Quantity 5 kg
Ignition Residue 0.001
Loss on Drying 0.01
CAS Max % 1
Percent Purity 98 to 102%