Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tripeptidyl-Peptidase I/TPP1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP136742
Description
Tripeptidyl-Peptidase I/TPP1 Polyclonal specifically detects Tripeptidyl-Peptidase I/TPP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
Tripeptidyl-Peptidase I/TPP1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Cell growth-inhibiting gene 1 protein, ceroid-lipofuscinosis, neuronal 2, late infantile (Jansky-Bielschowsky disease), CLN2EC 3.4.14.9, growth-inhibiting protein 1, LPIC, lysosomal pepstatin insensitive protease, Lysosomal pepstatin-insensitive protease, MGC21297, TPP-1, TPP-I, Tripeptidyl aminopeptidase, tripeptidyl peptidase I, tripeptidyl-peptidase 1, Tripeptidyl-peptidase I | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Dog and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Peptide ELISA Detection limit 1:4000 | |
NP_000382.3 | |
TPP1 | |
Peptide with sequence C-TPSVIRKRYNLTSQD corresponding to internal region according to NP_000382.3. | |
0.1 mg | |
Neuroscience | |
1200 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction