Learn More
Tripeptidyl-Peptidase I/TPP1 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0584-9V1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212194AF350
Catalog No. NP212194A30
Add to cart
Description
Description
Tripeptidyl-Peptidase I/TPP1 Monoclonal antibody specifically detects Tripeptidyl-Peptidase I/TPP1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Tripeptidyl-Peptidase I/TPP1
MM0584-9V1
Western Blot
Cell growth-inhibiting gene 1 protein, ceroid-lipofuscinosis, neuronal 2, late infantile (Jansky-Bielschowsky disease), CLN2EC 3.4.14.9, growth-inhibiting protein 1, LPIC, lysosomal pepstatin insensitive protease, Lysosomal pepstatin-insensitive protease, MGC21297, TPP-1, TPP-I, Tripeptidyl aminopeptidase, tripeptidyl peptidase I, tripeptidyl-peptidase 1, Tripeptidyl-peptidase I
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
TPP1
Human TPP1
0.1 mL
Primary
1200