Learn More
Alfa Aesar™ Tripeptidyl Peptidase Inhibitor
Manufacturer:  Alfa Aesar™ J64760MA
 View more versions of this product
Catalog No. AAJ64760MA
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II
Specifications
Specifications
10mg
−30° to −10°C
A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II
C16H22ClN3O3
H-AAF-CMK; TPPII Inhibitor
Signal Transduction Reagent
Off-white
Solid
184901-82-4
453.90g/mol
Light Sensitive