Alfa Aesar™ Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride, 98%

Manufacturer: Alfa Aesar™ H5504003

Quantity 1g

 View more versions of this product

Catalog No. AAH5504003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99662-46-1
    Molecular Formula C24H21ClNP·HCl
    Formula Weight 426.33
    MDL Number MFCD00674065
    Chemical Name or Material Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride

    Specifications

    Formula Weight 426.33
    Molecular Formula C24H21ClNP·HCl
    MDL Number MFCD00674065
    CAS 99662-46-1
    Melting Point 249° to 254°C
    Quantity 1g
    Chemical Name or Material Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit