Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
99662-46-1
C24H21ClNP·ClH
426.32
JHQWTRCLYUFMSJ-UHFFFAOYSA-M
triphenyl 2-pyridylmethyl phosphonium chloride hydrochloride,triphenyl pyridin-2-ylmethyl phosphanium hydrochloride chloride,acmc-20ajcr,triphenyl 2-pyridylmethyl-phosphonium chloride hydrochloride,triphenyl 2-pyridylmethyl phosphonium ch loride hydrochloride,triphenyl pyridin-2-yl methyl phosphanium chloride-hydrogen chloride 1/1/1
16213046
triphenyl(pyridin-2-ylmethyl)phosphanium;chloride;hydrochloride
C1=CC=C(C=C1)[P+](CC2=CC=CC=N2)(C3=CC=CC=C3)C4=CC=CC=C4.Cl.[Cl-]
99662-46-1
C24H21ClNP·ClH
426.32
JHQWTRCLYUFMSJ-UHFFFAOYSA-M
triphenyl 2-pyridylmethyl phosphonium chloride hydrochloride,triphenyl pyridin-2-ylmethyl phosphanium hydrochloride chloride,acmc-20ajcr,triphenyl 2-pyridylmethyl-phosphonium chloride hydrochloride,triphenyl 2-pyridylmethyl phosphonium ch loride hydrochloride,triphenyl pyridin-2-yl methyl phosphanium chloride-hydrogen chloride 1/1/1
16213046
triphenyl(pyridin-2-ylmethyl)phosphanium;chloride;hydrochloride
C1=CC=C(C=C1)[P+](CC2=CC=CC=N2)(C3=CC=CC=C3)C4=CC=CC=C4.Cl.[Cl-]
Quantity 5g
Color White to Yellow
Melting Point 241.0°C to 244.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to max. 102.5% min. On dry basis (Argentometry)
Formula Weight 426.32
Physical Form Low Melting Solid
Percent Purity 98%
Chemical Name or Material Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning