Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyl Phosphite 97.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
101-02-0
C18H15O3P
310.289
MFCD00003032
HVLLSGMXQDNUAL-UHFFFAOYSA-N
phosphorous acid, triphenyl ester,advance tpp,triphenoxyphosphine,phenyl phosphite,stabilizer p 36,trifenoxyfosfin,trifenylfosfit,phosclere t 36,efed,mellite 310
7540
triphenyl phosphite
C1=CC=C(C=C1)OP(OC2=CC=CC=C2)OC3=CC=CC=C3
101-02-0
C18H15O3P
310.289
MFCD00003032
HVLLSGMXQDNUAL-UHFFFAOYSA-N
phosphorous acid, triphenyl ester,advance tpp,triphenoxyphosphine,phenyl phosphite,stabilizer p 36,trifenoxyfosfin,trifenylfosfit,phosclere t 36,efed,mellite 310
7540
triphenyl phosphite
C1=CC=C(C=C1)OP(OC2=CC=CC=C2)OC3=CC=CC=C3
Boiling Point 360°C
Quantity 25g
Color Yellow
UN Number 3082
Formula Weight 310.29
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material Triphenyl Phosphite

EINECSNumber : (3)-2501

RTECSNumber : TH1575000

TSCA : Yes