Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylacetic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
595-91-5
C20H16O2
288.34
MFCD00004185
DCYGAPKNVCQNOE-UHFFFAOYSA-N
triphenylacetic acid,acetic acid, triphenyl,triphenyl acetic acid,benzeneacetic acid, .alpha.,.alpha.-diphenyl,nsc 61,tri phenylacetic acid,pubchem9200,alpha-toluic acid, alpha,alpha-diphenyl,acmc-1arvb,maybridge1_006927
68992
2,2,2-triphenylacetic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C(=O)O
595-91-5
C20H16O2
288.34
MFCD00004185
DCYGAPKNVCQNOE-UHFFFAOYSA-N
triphenylacetic acid,acetic acid, triphenyl,triphenyl acetic acid,benzeneacetic acid, .alpha.,.alpha.-diphenyl,nsc 61,tri phenylacetic acid,pubchem9200,alpha-toluic acid, alpha,alpha-diphenyl,acmc-1arvb,maybridge1_006927
68992
2,2,2-triphenylacetic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C(=O)O
Melting Point 268°C to 269°C
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color White to Beige
Assay Percent Range 99%
Linear Formula (C6H5)3CCO2H
Beilstein 09, 712
Formula Weight 288.34
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Triphenylacetic acid, 99%
Show More Show Less

EINECSNumber : 209-873-4