Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylcarbenium hexachloroantimonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
50g
1586-91-0
C19H15Cl6Sb
577.789
GASWLBDVMZXHOP-UHFFFAOYSA-H
tritylium hexachloroantimonate,triphenylcarbenium hexachloroantimonate,trityl hexachloroantimonate,hexachlorostibanuide; triphenylmethylium
16688166
diphenylmethylbenzene;hexachloroantimony(1-)
C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3.Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl
1586-91-0
C19H15Cl6Sb
577.789
GASWLBDVMZXHOP-UHFFFAOYSA-H
tritylium hexachloroantimonate,triphenylcarbenium hexachloroantimonate,trityl hexachloroantimonate,hexachlorostibanuide; triphenylmethylium
16688166
diphenylmethylbenzene;hexachloroantimony(1-)
C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3.Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl
Elemental Analysis 97.5% min.
Color Yellow
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 98%
Linear Formula (C6H5)3CSbCl6
Formula Weight 577.79
Physical Form Powder
Chemical Name or Material Triphenylcarbenium hexachloroantimonate, 98%

MOISTURE SENSITIVE

EINECSNumber : 216-446-6

TSCA : TSCA