Alfa Aesar™ Triphenylcarbenium hexachloroantimonate, 99%

Manufacturer: Alfa Aesar™ B2503506

Quantity 5g

 View more versions of this product

Catalog No. AAB2503506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1586-91-0
    Molecular Formula C19H15Cl6Sb
    Linear Formula (C6H5)3CSbCl6
    Formula Weight 577.8
    MDL Number MFCD00013126
    UN Number UN3261
    Beilstein 3817547
    Synonym Tritylium hexachloroantimonate; Trityl hexachloroantimonate
    Chemical Name or Material Triphenylcarbenium hexachloroantimonate

    Specifications

    Formula Weight 577.8
    TSCA TSCA
    MDL Number MFCD00013126
    UN Number UN3261
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P273-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Quantity 5g
    Odor Odorless
    Molecular Formula C19H15Cl6Sb
    Linear Formula (C6H5)3CSbCl6
    CAS 1586-91-0
    Beilstein 3817547
    Synonym Tritylium hexachloroantimonate; Trityl hexachloroantimonate
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Melting Point ≈220°C (dec.)
    Chemical Name or Material Triphenylcarbenium hexachloroantimonate
    Assay 99%

    GHS H Statement
    H314-H318-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P280-P273-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit