Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triphenylcarbenium hexachloroantimonate, 99%

Click to view available options
Quantity:
5g
25g
100g
1586-91-0
C19H15Cl6Sb
577.789
MFCD00013126
GASWLBDVMZXHOP-UHFFFAOYSA-H
tritylium hexachloroantimonate,triphenylcarbenium hexachloroantimonate,trityl hexachloroantimonate,hexachlorostibanuide; triphenylmethylium
16688166
diphenylmethylbenzene;hexachloroantimony(1-)
C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3.Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl
1586-91-0
C19H15Cl6Sb
577.789
MFCD00013126
GASWLBDVMZXHOP-UHFFFAOYSA-H
tritylium hexachloroantimonate,triphenylcarbenium hexachloroantimonate,trityl hexachloroantimonate,hexachlorostibanuide; triphenylmethylium
16688166
diphenylmethylbenzene;hexachloroantimony(1-)
C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3.Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl
Melting Point ~220°C (decomposition)
Quantity 25g
Odor Odorless
Linear Formula (C6H5)3CSbCl6
UN Number UN3261
Beilstein 3817547
Formula Weight 577.8
Percent Purity 99%
Chemical Name or Material Triphenylcarbenium hexachloroantimonate

GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P280-P273-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA