Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylcarbenium pentachlorostannate, 98%, Acros Organics

Click to view available options
Quantity:
25g
15414-98-9
C19H15Cl5Sn
539.289
MFCD00000512
LJHTYNWDYRJYIH-UHFFFAOYSA-I
trityl pentachlorostannate,triphenylcarbenium pentachlorostannate,methylium, triphenyl-, pentachlorostannate 1-,methylium, triphenyl-, pentachlorostannate 1-1:1,pentachlorostannanuide; triphenylmethylium
16717610
diphenylmethylbenzene;pentachlorostannanuide
C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3.Cl[Sn-](Cl)(Cl)(Cl)Cl
15414-98-9
C19H15Cl5Sn
539.289
MFCD00000512
LJHTYNWDYRJYIH-UHFFFAOYSA-I
trityl pentachlorostannate,triphenylcarbenium pentachlorostannate,methylium, triphenyl-, pentachlorostannate 1-,methylium, triphenyl-, pentachlorostannate 1-1:1,pentachlorostannanuide; triphenylmethylium
16717610
diphenylmethylbenzene;pentachlorostannanuide
C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3.Cl[Sn-](Cl)(Cl)(Cl)Cl
Color Brown to Orange-Yellow
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (C6H5)3CSnCl5
Formula Weight 539.28
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Triphenylcarbenium pentachlorostannate, 98%

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger