Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylene 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
217-59-4
C18H12
228.29
MFCD00001108
SLGBZMMZGDRARJ-UHFFFAOYSA-N
9,10-benzophenanthrene,isochrysene,9,10-benzphenanthrene,benzo l phenanthrene,1,2,3,4-dibenznaphthalene,benzophenanthrene,1,2:3,4-dibenznaphthalene,ccris 1301,triphenylen-1-yl
9170
CHEBI:33080
triphenylene
C1=CC=C2C(=C1)C3=CC=CC=C3C4=CC=CC=C24
217-59-4
C18H12
228.29
MFCD00001108
SLGBZMMZGDRARJ-UHFFFAOYSA-N
9,10-benzophenanthrene,isochrysene,9,10-benzphenanthrene,benzo l phenanthrene,1,2,3,4-dibenznaphthalene,benzophenanthrene,1,2:3,4-dibenznaphthalene,ccris 1301,triphenylen-1-yl
9170
CHEBI:33080
triphenylene
C1=CC=C2C(=C1)C3=CC=CC=C3C4=CC=CC=C24
Melting Point 196.0°C to 200.0°C
Color White to Beige
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Boiling Point 438.0°C
Assay Percent Range 98%
Beilstein 05, 720
Merck Index 15, 9913
Formula Weight 228.29
Physical Form Crystalline Needles
Percent Purity 98%
Chemical Name or Material Triphenylene, 98%
Show More Show Less

EINECSNumber : 205-922-9

RTECSNumber : YK2925000