Alfa Aesar™ Triphenylmethane, 98%

Manufacturer: Alfa Aesar™ A1171036

Quantity 500g

 View more versions of this product

Catalog No. AAA1171036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 519-73-3
    Molecular Formula (C6H5)3CH
    Formula Weight 244.34
    MDL Number MFCD00004763
    Beilstein 1909753
    Merck Index 14,9741
    Synonym Tritan
    Chemical Name or Material Triphenylmethane

    Specifications

    Formula Weight 244.34
    TSCA TSCA
    Molecular Formula (C6H5)3CH
    Synonym Tritan
    Boiling Point 358° to 359°C
    MDL Number MFCD00004763
    CAS 519-73-3
    Merck Index 14,9741
    Melting Point 92° to 94°C
    Beilstein 1909753
    Quantity 500g
    Chemical Name or Material Triphenylmethane
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit