Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylmethanol, 97%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Plastic bottle
76-84-6
C19H16O
260.32
LZTRCELOJRDYMQ-UHFFFAOYSA-N
triphenylcarbinol,trityl alcohol,tritanol,triphenylmethyl alcohol,triphenyl methanol,triphenyl carbinol,methanol, triphenyl,unii-u97q0ou9kb,triphenyl-methanol,benzenemethanol, .alpha.,.alpha.-diphenyl
6457
triphenylmethanol
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O
76-84-6
C19H16O
260.32
LZTRCELOJRDYMQ-UHFFFAOYSA-N
triphenylcarbinol,trityl alcohol,tritanol,triphenylmethyl alcohol,triphenyl methanol,triphenyl carbinol,methanol, triphenyl,unii-u97q0ou9kb,triphenyl-methanol,benzenemethanol, .alpha.,.alpha.-diphenyl
6457
triphenylmethanol
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O
Boiling Point 360°C
Packaging Plastic bottle
Quantity 50g
Melting Point 160°C to 164°C
Infrared Spectrum Authentic
Color White to Yellow
Assay Percent Range 97%
Linear Formula (C6H5)3COH
Beilstein 06,713
Merck Index 15,9912
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. h2so4 and glacial acetic acid,soluble in alcohol,ether and benzene
Formula Weight 260.32
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material Triphenylmethanol
Show More Show Less

EINECSNumber : 200-988-5